(R)-(+)-Limonene ≥97%

Supplier: Matrix Scientific

084604-50G
102842-866EA 38.46 USD
102842-866
(R)-(+)-Limonene ≥97%
(R)-(+)-Limonene

Hazard: Irritant

Formula: C₁₀H₁₆
MDL Number: MFCD00062991
CAS Number: 5989-27-5
UN: 2052
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR