4-Methoxyphenylphosphonic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Phosphonylanisole

084474-1G
102842-506EA 383.08 USD
102842-506
4-Methoxyphenylphosphonic acid ≥97%
4-Methoxyphenylphosphonic acid

Hazard: Irritant

Formula: C₇H₉O₄P
MW: 188.12 g/mol
MDL Number: MFCD00013948
CAS Number: 21778-19-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR