1-(Chloromethyl)cyclohexanecarbonitrile ≥95%

Supplier: Matrix Scientific

084384-1G
102842-262EA 132.31 USD
102842-262
1-(Chloromethyl)cyclohexanecarbonitrile ≥95%
1-(Chloromethyl)cyclohexanecarbonitrile

Hazard: Irritant

Formula: C₈H₁₂ClN
MW: 157.64 g/mol
MDL Number: MFCD01570824
CAS Number: 112905-95-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR