5-(Chloromethyl)-1-methyl-3-phenyl-1H-pyrazole ≥97%

Supplier: Matrix Scientific

084360-1G
102842-196EA 949.23 USD
102842-196
5-(Chloromethyl)-1-methyl-3-phenyl-1H-pyrazole ≥97%
5-(Chloromethyl)-1-methyl-3-phenyl-1H-pyrazole

Hazard: Irritant

Formula: C₁₁H₁₁ClN₂
MDL Number: MFCD08271944
CAS Number: 869901-14-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR