tert-Butyl (2S)-4,4-diethoxy-2-(hydroxymethyl)piperidine-1-carboxylate ≥97%

Supplier: Matrix Scientific

084249-1G
102841-892EA 1372.31 USD
102841-892
tert-Butyl (2S)-4,4-diethoxy-2-(hydroxymethyl)piperidine-1-carboxylate ≥97%
tert-Butyl (2S)-4,4-diethoxy-2-(hydroxymethyl)piperidine-1-carboxylate

Hazard: Irritant

Formula: C₁₅H₂₉NO₅
MDL Number: MFCD08056332
CAS Number: 1212360-70-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR