2,7-Dichloroimidazo[1,2-a]pyridine ≥97%

Supplier: Matrix Scientific

084131-1G
102841-586EA 636.92 USD
102841-586
2,7-Dichloroimidazo[1,2-a]pyridine ≥97%
2,7-Dichloroimidazo[1,2-a]pyridine

Hazard: Irritant

Formula: C₇H₄Cl₂N₂
MDL Number: MFCD06739216
CAS Number: 190074-50-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR