2-Chloroimidazole ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Chloro-1H-imidazole

083942-1G
102841-136EA 750.77 USD
102841-136
2-Chloroimidazole ≥97%
2-Chloroimidazole

Hazard: Irritant

Formula: C₃H₃ClN₂
MW: 102.52
MDL Number: MFCD02179530
CAS Number: 16265-04-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR