4-Bromo-3-methylanisole ≥97%

Supplier: Matrix Scientific

Synonyms: 1-Bromo-4-methoxy-2-methylbenzene, 2-Bromo-5-methoxytoluene

083807-50G
102840-784EA 95.38 USD
102840-784
4-Bromo-3-methylanisole ≥97%
4-Bromo-3-methylanisole

Hazard: Irritant

Formula: C₈H₉BrO
MDL Number: MFCD00060687
CAS Number: 27060-75-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR