Ethyl-3,5-dichloroimidazo[1,2-a]pyridine-2-carboxylate ≥97%

Supplier: Matrix Scientific

083648-1G
102840-316EA 849.23 USD
102840-316
Ethyl-3,5-dichloroimidazo[1,2-a]pyridine-2-carboxylate ≥97%
Ethyl-3,5-dichloroimidazo[1,2-a]pyridine-2-carboxylate

Hazard: Irritant

Formula: C₁₀H₈Cl₂N₂O₂
MDL Number: MFCD09842623
CAS Number: 1000018-01-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR