Ethyl-2-phenylacrylate ≥97%

Supplier: Matrix Scientific

Synonyms: Ethyl 2-phenylacrylate

083239-1G
102839-254EA 384.62 USD
102839-254
Ethyl-2-phenylacrylate ≥97%
Ethyl-2-phenylacrylate

Hazard: Irritant

Formula: C₁₁H₁₂O₂
Boiling Pt: 123…125 °C
MDL Number: MFCD00026862
CAS Number: 22286-82-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR