Mercaptoacetic acid (thioglycolic acid) ≥97%

Supplier: Matrix Scientific

Synonyms: Mercaptoacetic acid

083060-10G 083060-50G
102838-818EA 306.15 USD
102838-818 102838-820
Mercaptoacetic acid (thioglycolic acid) ≥97%
Mercaptoacetic acid (Thioglycolic acid)

Hazard: Corrosive/Air Sensitive

Formula: HSCH₂COOH
MW: 92.12 g/mol
Boiling Pt: 96
Melting Pt: -16.5
Density: 1.295
Flash Pt: 119 °C
MDL Number: MFCD00004876
CAS Number: 68-11-1
UN: 1940
ADR: 8 II
Merck Index: 13,09410
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=45179825145ec44f2e058197a0ee54845ae7c750&vid=51e43f5b69b880b196570b3065f9b4d8db7336d1&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR