Benzoin hemisuccinate ≥97%

Supplier: Matrix Scientific

083025-1G
102838-690EA 638.46 USD
102838-690
Benzoin hemisuccinate ≥97%
Benzoin hemisuccinate

Hazard: Irritant

Formula: C₁₈H₁₈O₄
MDL Number: MFCD01313222
CAS Number: 306935-85-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR