2-Methylindolizine-6-carboxylic acid ≥97%

Supplier: Matrix Scientific

082955-1G
102838-516EA 1530.77 USD
102838-516
2-Methylindolizine-6-carboxylic acid ≥97%
2-Methylindolizine-6-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₉NO₂
MDL Number: MFCD08056317
CAS Number: 256935-78-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR