1-Morpholin-4-ylpyrrolidine-3-carboxylic acid ≥97%

Supplier: Matrix Scientific

082799-1G
102838-146EA 806.15 USD
102838-146
1-Morpholin-4-ylpyrrolidine-3-carboxylic acid ≥97%
1-Morpholin-4-ylpyrrolidine-3-carboxylic acid

Hazard: Irritant

Formula: C₉H₁₆N₂O₃
MW: 200.24 g/mol
MDL Number: MFCD09258860
CAS Number: 1086374-82-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR