5-Chloro-1-methyl-1H-pyrazole-3-carboxylic acid ≥97%

Supplier: Matrix Scientific

082628-1G
102837-754EA 1032.31 USD
102837-754
5-Chloro-1-methyl-1H-pyrazole-3-carboxylic acid ≥97%
5-Chloro-1-methyl-1H-pyrazole-3-carboxylic acid

Hazard: Irritant

Formula: C₅H₅ClN₂O₂
MW: 160.56 g/mol
MDL Number: MFCD00106211
CAS Number: 1173246-76-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR