Nicotinic acid (Niacin) ≥97%
Supplier: Matrix Scientific
Synonyms:
3-Pyridinecarboxylic acid, Pyridine-3-carboxylic acid, 3-Picolinic acid, Niacin, Nicotinic acid, Vitamin B3
082487-50G
102837-444EA
61.54
USD
102837-444
Nicotinic acid (Niacin) ≥97%
Nicotinic acid (Niacin)
Purity: 97%
CAS: 59-67-6
MDL Number: MFCD00006391
Molecular Formula: C6H5NO2
Molecular Weight: 123,11
Hazard: Irritant
Formula:
C₆H₅NO₂ MW: 123.11 g/mol |
MDL Number:
MFCD00006391 CAS Number: 59-67-6 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...