4-Amino-3-(trifluoromethoxy)benzoic acid ≥97%

Supplier: Matrix Scientific

082275-1G 082275-10G
102836-980EA 200 USD
102836-980 102836-982
4-Amino-3-(trifluoromethoxy)benzoic acid ≥97%
4-Amino-3-(trifluoromethoxy)benzoic acid

Hazard: Irritant

Formula: C₈H₆F₃NO₃
MDL Number: MFCD00205143
CAS Number: 175278-22-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR