3-(1H-Imidazol-4-yl)benzoic acid ≥95%

Supplier: Matrix Scientific

082256-1G
102836-936EA 1115.38 USD
102836-936
3-(1H-Imidazol-4-yl)benzoic acid ≥95%
3-(1H-Imidazol-4-yl)benzoic acid

Hazard: Irritant

Formula: C₁₀H₈N₂O₂
MDL Number: MFCD08056299
CAS Number: 912569-71-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR