2-(Phenoxymethyl)benzoic acid ≥97%

Supplier: Matrix Scientific

082205-1G 082205-10G
102836-808EA 101.54 USD
102836-808 102836-810
2-(Phenoxymethyl)benzoic acid ≥97%
2-(Phenoxymethyl)benzoic acid

Hazard: Irritant

Formula: C₁₄H₁₂O₃
MDL Number: MFCD00020294
CAS Number: 724-98-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR