4-Amino-3-iodobenzoic acid ≥97%

Supplier: Matrix Scientific

082021-1G
102836-348EA 153.85 USD
102836-348
4-Amino-3-iodobenzoic acid ≥97%
4-Amino-3-iodobenzoic acid

Hazard: Irritant

Formula: C₇H₆INO₂
MW: 263.03 g/mol
MDL Number: MFCD01660550
CAS Number: 2122-63-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR