Phenylboronic acid neopentyl glycol ester ≥97%

Supplier: Matrix Scientific

Synonyms: 5,5-Dimethyl-2-phenyl-1,3,2-dioxaborinane

081990-1G 081990-10G
102836-274EA 76.92 USD
102836-274 102836-276
Phenylboronic acid neopentyl glycol ester ≥97%
Phenylboronic acid neopentyl glycol ester

Hazard: Irritant

Formula: C₁₁H₁₅BO₂
MDL Number: MFCD05663851
CAS Number: 5123-13-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR