(4-Methyl-2-phenyl-1,3-thiazol-5-yl)methylamine ≥97%

Supplier: Matrix Scientific

081915-1G
102836-110EA 553.85 USD
102836-110
(4-Methyl-2-phenyl-1,3-thiazol-5-yl)methylamine ≥97%
(4-Methyl-2-phenyl-1,3-thiazol-5-yl)methylamine

Hazard: Irritant

Formula: C₁₁H₁₂N₂S
MDL Number: MFCD05865105
CAS Number: 165735-95-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR