N-Methyl-N-[2-(trifluoromethyl)benzyl]amine ≥97%

Supplier: Matrix Scientific

Synonyms: N-Methyl-2-trifluoromethylbenzylamine

081800-1G
102835-850EA 636.92 USD
102835-850
N-Methyl-N-[2-(trifluoromethyl)benzyl]amine ≥97%
N-Methyl-N-[2-(trifluoromethyl)benzyl]amine

Hazard: Irritant

Formula: C₉H₁₀F₃N
MDL Number: MFCD02689994
CAS Number: 296276-41-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR