3-(Bromomethyl)benzaldehyde ≥97%

Supplier: Matrix Scientific

081542-1G
102835-224EA 496.92 USD
102835-224
3-(Bromomethyl)benzaldehyde ≥97%
3-(Bromomethyl)benzaldehyde

Hazard: Corrosive/Stored under Argon/Light Sensitive/Air Sensitive

Formula: C₈H₇BrO
Melting Pt: 49…51 °C
MDL Number: MFCD06797497
CAS Number: 82072-23-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR