5-Pyridin-4-yl-1,2,4-oxadiazol-3-amine ≥97%

Supplier: Matrix Scientific

081377-1G
102834-746EA 461.54 USD
102834-746
5-Pyridin-4-yl-1,2,4-oxadiazol-3-amine ≥97%
5-Pyridin-4-yl-1,2,4-oxadiazol-3-amine

Hazard: Irritant

Formula: C₇H₆N₄O
MDL Number: MFCD09864897
CAS Number: 23275-49-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR