3-Aminopyridine-2,4-diol ≥97%

Supplier: Matrix Scientific

081265-1G
102834-470EA 596.92 USD
102834-470
3-Aminopyridine-2,4-diol ≥97%
3-Aminopyridine-2,4-diol

Hazard: Irritant

Formula: C₅H₆N₂O₂
MDL Number: MFCD09991480
CAS Number: 103792-82-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR