6-(Tetrahydro-2H-pyran-4-ylmethoxy)pyridin-3-amine ≥97%

Supplier: Matrix Scientific

081238-1G
102834-406EA 670.77 USD
102834-406
6-(Tetrahydro-2H-pyran-4-ylmethoxy)pyridin-3-amine ≥97%
6-(Tetrahydro-2H-pyran-4-ylmethoxy)pyridin-3-amine

Hazard: Irritant

Formula: C₁₁H₁₆N₂O₂
MDL Number: MFCD12026336
CAS Number: 1180133-66-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR