Cytosine

Supplier: Matrix Scientific

Synonyms: 6-Aminopyrimidin-2(1H)-one, 4-Amino-2-pyrimidinol

080690-100G
102833-972EA 87.69 USD
102833-972
Cytosine
Cytosine

Hazard: Irritant

Formula: C₄H₅N₃O
MW: 111.1 g/mol
MDL Number: MFCD00006034
CAS Number: 71-30-7
Merck Index: 13,02824

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR