2,3-Dihydro-1H-inden-2-amine hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Aminoindan hydrochloride, 2-Indanylamine hydrochloride

075887-5G 075887-25G 075887-100G
102543-474EA 73.85 USD
102543-474 102833-882 102833-884
2,3-Dihydro-1H-inden-2-amine hydrochloride ≥95%
2,3-Dihydro-1H-inden-2-amine hydrochloride

Hazard: Irritant

Formula: C₉H₁₂ClN
MDL Number: MFCD00012549
CAS Number: 2338-18-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR