2',3'-O-Isopropylideneadenosine ≥95%

Supplier: Matrix Scientific

075482-100G 075482-25G
102541-562EA 158.46 USD
102541-562 102833-834
2',3'-O-Isopropylideneadenosine ≥95%
2',3'-O-Isopropylideneadenosine

Hazard: Irritant

Formula: C₁₃H₁₇N₅O₄
Melting Pt: 221…222 °C
MDL Number: MFCD00005756
CAS Number: 362-75-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR