D-Cycloserine ≥95%

Supplier: Matrix Scientific

Synonyms: D-(+)-Cycloserine, Cycloserine, D(+)-Cycloserine, (R)-4-Aminoisoxazolidin-3-one, D-4-Aminoisoxazolidin-3-one, (R)-(+)-Cycloserine

072929-5G 072929-25G
102527-496EA 138.46 USD
102527-496 102833-574
D-Cycloserine ≥95%
D-Cycloserine

Hazard: Irritant

Formula: C₃H₆N₂O₂
MW: 102.09 g/mol
MDL Number: MFCD00005353
CAS Number: 68-41-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR