4,6-Dimethoxyindole

Supplier: Matrix Scientific

064205-1G 064205-5G
102833-356EA 526.15 USD
102833-356 102833-358
4,6-Dimethoxyindole
4,6-Dimethoxyindole

Hazard: Irritant

Formula: C₁₀H₁₁NO₂
MW: 177.2
MDL Number: MFCD00128213
CAS Number: 23659-87-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR