Cytarabine ≥95%

Supplier: Matrix Scientific

Synonyms: Cytosine beta-D-Arabinofuranoside, 1-β-D-Arabinofuranosylcytosine, Arabinocytidine, Ara-C, 4-Amino-1-((2R,3S,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one, Cytosine arabinoside, 4-Amino-1-beta-D-arabinofuranosyl-2(1H)-pyrimidinone

047887-25G
102833-152EA 98.46 USD
102833-152
Cytarabine ≥95%
Cytarabine

Purity: 95% CAS: 69-74-9 MDL Number: MFCD00066487 Molecular Formula: C9H13N3O5 Molecular Weight: 243,22 Hazard: Irritant

Formula: C₉H₁₃N₃O₅
MW: 243.22 g/mol
Melting Pt: 197…198 °C
MDL Number: MFCD00066487
CAS Number: 147-94-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR