β-Alanine-tert-butylester hydrochloride

Supplier: Matrix Scientific

037595-5G
102833-078EA 30.77 USD
102833-078
β-Alanine-tert-butylester hydrochloride
beta-Alanine-tert-butylester hydrochloride

Hazard: Irritant

Formula: C₇H₁₆ClNO₂
MDL Number: MFCD00058258
CAS Number: 58620-93-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR