Human Recombinant Neuritin (from E. coli)
Supplier: BioVendor
RD172358100
102830-656EA
583.35
USD
102830-656
Human Recombinant Neuritin (from E. coli)
Proteins and Peptides
Neuritin was identified as an important neurotrophin expressed in the developing nervous system. This protein plays multiple roles in the process of neural development, synaptic plasticity, and synaptic maturation.
Sequence: MKHHHHHHAS AGKCDAVFKG FSDCLLKLGD SMANYPQGLD DKTNIKTVCT YWEDFHSCTV TALTDCQEGA KDMWDKLRKE SKNLNIQGSL FELCGSGNG
Other names: NRN1, NRN
Ordering information: For research use only.
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...