CLL Serum, Charles River Laboratories Cell Solutions
Supplier: Charles River Laboratories Cell Solutions
PB007C-1-CLL
102830-082EA 297.69 USD
102830-082
CLL Serum, Charles River Laboratories Cell Solutions
Sera (Serum)
Human disease state peripheral blood serum is collected in HemaCare's FDA-registered collection centers or through our hospital or physician office network from IRB consented human volunteer donors
Serological and Infectious disease testing are performed with each donation. Customers should handle product as if potentially infectious following biosafety level 2 procedures.
Serum is collected from peripheral blood in a plain or Serum Separator Tube.