Human Bone Marrow Mesenchymal Stem Cells (MSC), Charles River Laboratories Cell Solutions
Supplier: Charles River Laboratories Cell Solutions
BM012C-1
102830-012EA 1653.85 USD
102830-012
Human Bone Marrow Mesenchymal Stem Cells (MSC), Charles River Laboratories Cell Solutions
Cells
Human bone marrow mesenchymal stem cells (MSC) are isolated by adherence from bone marrow mononuclear cells (MNCs), isolated using a density gradient
Adherent cells were cultured in MEM-Alpha, supplemented with L-Glutamine and Human Platelet Lysate. Human bone marrow is collected from healthy human volunteer donors under IRB approved informed consent via our qualified hospital and physician network.