QC Colloidal Coomassie Stain, Bio-Rad
Supplier: BIO-RAD LABORATORIES MS
1610803
102827-652EA 192.31 USD
102827-652
QC Colloidal Coomassie Stain, Bio-Rad
Stains Electrophoresis Stains
Ready-to-use, non-hazardous colloidal Coomassie G-250 stain for protein polyacrylamide gels. Use QC Colloidal Coomassie to stain proteins in SDS-PAGE gels without using methanol for fixing.
  • Low background, high sensitivity, superior reproducibility
  • Environmentally friendly formulation — no addition of methanol or acetic acid required; eliminates the need for hazardous waste disposal
  • Flexible staining and destaining times — from 1 hour to overnight
  • No alcohol addition or dilution steps when staining polyacrylamide gels
  • One-part, ready-to-use colloidal Coomassie stain