Anti-RBM15 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Bioss

BS-12402R-FITC
10281-406EA 823.27 USD
10281-406
Anti-RBM15 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
RBM15 is a 977 amino acid protein that localizes to the nucleus and contains one SPOC domain and three RRM domains. Expressed as multiple alternatively spliced isoforms, RBM15 interacts with Epstein-Barr (EBV) viral proteins and is thought to be involved in the regulation of Hox genes, possibly via interactions with RNA and spliceosome components. RBM15 is subject to post-translational phosphorylation, probably by ATM or ATR. Chromosomal aberrations involving the RBM15 gene, which localizes to human chromosome 1, may be associated with the development of acute megakaryoblastic leukemia.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RBM15
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR