Nestlets™ Nesting Material, Ancare
Supplier: Ancare
NES3600
10279-140CS 1118.23 USD
10279-140
Nestlets™ Nesting Material, Ancare
Animal Supplies Animal Bedding
Ideal for use with mice, hamsters, gerbils, rats, rabbits, and other small animals.
  • Clean, dust-free and autoclavable
  • Made of sterilized pulped virgin cotton fiber
  • Chemically inert odorless and noningestible
Available as 30.5×30.5 cm (12×12") sheets. Each package contains enough sheets to produce 3600 two-inch square Nestlets™.