Hydrogen chloride 0.01 N in methanol

Supplier: AQUA SOLUTIONS MS
Danger

4095-4LC 4095-4L
102777-940EA 211.92 USD
102777-940 103194-038
Hydrogen chloride 0.01 N in methanol
Hydrogen chloride
Formula: HCl
MDL Number: MFCD00011324
CAS Number: 7647-01-0
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Actual Normality 0.0095 - 0.0105 N


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR