4-Aminophthalic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

AC38224-0250
102774-524EA 49.9 USD
102774-524 102776-196 102777-552
4-Aminophthalic acid 97%
4-Aminophthalic acid
Formula: C₈H₇NO₄
MW: 181.15
Melting Pt: 344 °C
Storage Temperature: Ambient
MDL Number: MFCD00013985
CAS Number: 5434-21-9

Order Now

Specification Test Results

Appearance Beige Powder
Infrared spectrum Conforms
Titration with NaOH ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR