4-Iodophenylhydrazine 95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (4-Iodophenyl)hydrazine

AC39905-0050
102775-292EA 82 USD
102775-292 102776-950
4-Iodophenylhydrazine 95%
4-Iodophenylhydrazine
Formula: C₆H₇IN₂
MW: 234.04 g/mol
Melting Pt: 101…102 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00052688
CAS Number: 13116-27-3
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to orange to brown Crystalline powder
Infrared spectrum Conforms
Melting point 99 to 106 °C
NMR ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR