3-Methoxybenzenesulfonyl chloride 96%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3-Methoxybenzene-1-sulfonyl chloride, 3-Methoxybenzene-1-sulphonyl chloride

AC39852-0050
102775-214EA 78.9 USD
102775-214 102776-896
3-Methoxybenzenesulfonyl chloride 96%
3-Methoxybenzenesulphonyl chloride
Formula: C₇H₇ClO₃S
MW: 206.65 g/mol
Boiling Pt: 86…98 °C (1 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD01318155
CAS Number: 10130-74-2
UN: 3261
ADR: 8 II

Order Now

Specification Test Results

Appearance Light yellow to light brown Liquid or low melting solid
Infrared spectrum Conforms
GC ≥95 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR