Ethyl-4-methyl-4-pentenoate 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ethyl 4-methylpent-4-enoate

AC42737-0010
102774-954EA 75.92 USD
102774-954 102776-612
Ethyl-4-methyl-4-pentenoate 95%
Ethyl-4-methyl-4-pentenoate

Appearance: Clear colorless Liquid

Formula: C₈H₁₄O₂
MW: 142.198 g/mol
Boiling Pt: 85 °C (20 mmHg)
Density: 0.891 g/cm³
Flash Pt: 47 °C
Storage Temperature: Ambient
MDL Number: MFCD00075160
CAS Number: 4911-54-0
UN: 3272
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
GC ≥94 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR