Tetrachlorosilane 1M in methylene chloride, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Silicon tetrachloride, Silicon (IV) chloride

AC42876-1000
102774-690EA 57.8 USD
102774-690
Tetrachlorosilane 1M in methylene chloride, AcroSeal™
Tetrachlorosilane
Formula: Cl₄Si
MW: 169.9 g/mol
Boiling Pt: 57.6 °C (1013 hPa)
Melting Pt: –70 °C
Density: 1.34 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011229
CAS Number: 10026-04-7
UN: 2922
ADR: 8,I

Order Now

Specification Test Results

Appearance Clear colorless Solution
Strength 1 M exact strength on the certificate of analysis

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR