Peak ABC Loupe, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
68160
102773-570EA 775.66 USD
102773-570
Peak ABC Loupe, Electron Microscopy Sciences
Magnifiers
The Peak ABC Loupe is a precision folding loupe, consisting of A lens (12X), B lens (3X) and C lens (6X).
With these three lenses combined, seven kinds of magnifying power are available. The lens is made from high quality glass optics and the body is made of aluminum. The magnification desired will be available from the combination table shown on the back of the loupe.