Anti-DDX3X Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss

BS-12190R-CY7
10275-928EA 823.29 USD
10275-928
Anti-DDX3X Rabbit Polyclonal Antibody (Cy7®)
Antibodies
DDX3 is involved in RNA metabolism. Two DDX3 paralogs are found in humans; DDX3X is encoded by a gene found on the X chromosome while DDX3Y is encoded by a gene on the Y chromosome. DDX3Y is exclusively expressed in testis and is required for normal spermatogenesis. DDX3X is ubiquitously expressed and predominantly localizes to the nuclear speckles, participating in RNA splicing, transcription, translation initiation, mRNA transport and cell cycle regulation. DDX3X also partakes in HIV-1 replication and hepatitis C viral infections.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DDX3X
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR