Sheath Fluid (30X), Tonbo Biosciences
Supplier: Tonbo Biosciences
TNB-4600-L600
10274-086EA 49.5 USD
10274-086
Sheath Fluid (30X), Tonbo Biosciences
Buffers Assay Buffers
Tonbo Biosciences Sheath Fluid (30X) is a pre-filtered, pH balanced phosphate buffered saline solution that can be used for transporting particles though any flow cytometer.
This concentrated formula is easily prepared, uses less storage space and costs less to ship than conventional 20 L containers. Each bottle provides sufficient concentrate to prepare 20 liters of 1x flow cytometer sheath fluid.