Anti-BCHE Rabbit Polyclonal Antibody

Supplier: Proteintech
23854-1-AP-150UL
10273-138EA 462.47 USD
10273-138
Anti-BCHE Rabbit Polyclonal Antibody
Antibodies
The BCHE antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human BCHE. This antibody recognizes human antigen. The BCHE antibody has been validated for the following applications: ELISA, WB, IHC, IF analysis.

Type: Primary
Antigen: BCHE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR